Please login to your online account to display your discounted pricing

2-Bromophenyl acetate, 98%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2-Bromophenyl acetate
Assay Percent Range 98%
Molecular Formula C8H7BrO2
Formula Weight 215.05
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC452750010

  SDS
thermo fisher scientific
AC452750010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC452750050

  SDS
thermo fisher scientific
AC452750050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-Bromophenyl acetate
CAS 1829-37-4
Quantity 1g
Assay Percent Range 98%
Molecular Formula C8H7BrO2
Formula Weight 215.05