missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromophenyl acetate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC452750050

 View more versions of this product

Catalog No. AC452750050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Bromophenyl acetate
1829-37-4
C8H7BrO2
BEHBHYYPTOHUHX-UHFFFAOYSA-N
(2-bromophenyl) acetate
74586
98%
5g
97.5% min. (GC)
1-acetoxy-2-bromobenzene, 2-bromo-phenol acetate, 2-bromophenol acetate, 2-bromophenyl acetate, o-acetoxybromobenzene, o-bromophenyl acetate, phenol, 2-bromo-, acetate, phenol, o-bromo-, acetate, pubchem3055
CC(=O)OC1=CC=CC=C1Br
215.05
215.05

Chemical Identifiers

1829-37-4
215.05
1-acetoxy-2-bromobenzene, 2-bromo-phenol acetate, 2-bromophenol acetate, 2-bromophenyl acetate, o-acetoxybromobenzene, o-bromophenyl acetate, phenol, 2-bromo-, acetate, phenol, o-bromo-, acetate, pubchem3055
(2-bromophenyl) acetate
C8H7BrO2
BEHBHYYPTOHUHX-UHFFFAOYSA-N
74586
CC(=O)OC1=CC=CC=C1Br
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful if inhaled.

GHS Signal Word: Warning

EINECSNumber : 217-380-

SDS Documents