Learn More
2-Bromophenol, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 157360100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Bromophenol | |
95-56-7 | |
100.0 | |
195°C | |
Authentic | |
Glass bottle | |
1.5880 to 1.59 | |
MFCD00002146 | |
06, 197 | |
15, 1439 | |
Solubility in water: soluble in water. Other solubilities: miscible with chloroform and ether | |
C1=CC=C(C(=C1)O)Br | |
173.01 | |
173.01 |
98% | |
97.5 | |
1.4900g/mL | |
42°C | |
97.5% min. (GC) | |
C6H5BrO | |
BrC6H4OH | |
10mL | |
1.49 | |
o-bromophenol, phenol, 2-bromo, bromophenol, 2-bromfenol, 2-bromo phenol, 2-bromo-phenol, phenol, o-bromo, o-bromo-phenol, phenol, bromo, 2-bromfenol czech | |
VADKRMSMGWJZCF-UHFFFAOYSA-N | |
2-bromophenol | |
7244 | |
98% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Very toxic to aquatic life.
Flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 202-432-7
RTECSNumber : SJ7875000
TSCA : TSCA
RUO â Research Use Only