Learn More
2-Bromophenethyl alcohol, 99%, ACROS Organics™
$170.96 - $170.96
Chemical Identifiers
CAS | 1074-16-4 |
---|---|
Molecular Formula | C8H9BrO |
Molecular Weight (g/mol) | 201.06 |
MDL Number | MFCD00093566 |
InChI Key | ADLOWZRDUHSVRU-UHFFFAOYSA-N |
Synonym | 2-2-bromophenyl ethan-1-ol, 2-2-bromophenyl ethanol, 2-2-bromophenyl ethyl alcohol, 2-bromobenzeneethanol, 2-bromophenethyl alcohol, 2-bromophenethylalcohol, 2-bromophenylethanol, benzeneethanol, 2-bromo, o-bromophenethyl alcohol, pubchem16178 |
PubChem CID | 2734089 |
IUPAC Name | 2-(2-bromophenyl)ethanol |
SMILES | C1=CC=C(C(=C1)CCO)Br |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC350550010
|
Thermo Fisher Scientific
350550010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC350550050
|
Thermo Fisher Scientific
350550050 |
5g | Glass bottle |
Each for $170.96
|
|
Chemical Identifiers
1074-16-4 | |
201.06 | |
ADLOWZRDUHSVRU-UHFFFAOYSA-N | |
2734089 | |
C1=CC=C(C(=C1)CCO)Br |
C8H9BrO | |
MFCD00093566 | |
2-2-bromophenyl ethan-1-ol, 2-2-bromophenyl ethanol, 2-2-bromophenyl ethyl alcohol, 2-bromobenzeneethanol, 2-bromophenethyl alcohol, 2-bromophenethylalcohol, 2-bromophenylethanol, benzeneethanol, 2-bromo, o-bromophenethyl alcohol, pubchem16178 | |
2-(2-bromophenyl)ethanol |
Specifications
1.4830g/mL | |
Authentic | |
1.5755 to 1.5775 | |
97.0°C (0.7 mmHg) | |
1g | |
98.5 | |
98.5% min. (GC) | |
BrC6H4CH2CH2OH | |
2-2-bromophenyl ethan-1-ol, 2-2-bromophenyl ethanol, 2-2-bromophenyl ethyl alcohol, 2-bromobenzeneethanol, 2-bromophenethyl alcohol, 2-bromophenethylalcohol, 2-bromophenylethanol, benzeneethanol, 2-bromo, o-bromophenethyl alcohol, pubchem16178 | |
C1=CC=C(C(=C1)CCO)Br | |
201.06 | |
201.06 | |
99% |
>110°C | |
Glass bottle | |
1.483 | |
Yellow | |
1074-16-4 | |
100.0 | |
C8H9BrO | |
MFCD00093566 | |
ADLOWZRDUHSVRU-UHFFFAOYSA-N | |
2-(2-bromophenyl)ethanol | |
2734089 | |
Liquid | |
2-Bromophenethyl alcohol, 99% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
TSCA : TSCA