Learn More
2-Bromopentane, tech. 90%
CAS: 107-81-3 | C5H11Br | 151.047 g/mol
Supplier: Thermo Scientific Chemicals A1720536
Description
Used as a chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as a chemical intermediate.
Solubility
Not miscible or difficult to mix in water. Soluble in Dichloromethane, Ether, Ethyl Acetate, Hexane and Methanol
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
Specifications
| 2-Bromopentane | |
| -96°C | |
| 117°C to 118°C | |
| Characteristic | |
| 1.4403 | |
| 500 g | |
| 1718829 | |
| Not miscible or difficult to mix in water. Soluble in Dichloromethane,Ether,Ethyl Acetate,Hexane and Methanol | |
| CCCC(C)Br | |
| 151.047 | |
| 151.05 | |
| Technical |
| 107-81-3 | |
| 1.22 | |
| 20°C (68°F) | |
| C5H11Br | |
| MFCD00000160 | |
| UN2343 | |
| pentane, 2-bromo, 2-pentyl bromide, 1-methylbutyl bromide, +/--2-bromopentane, sec-amyl bromide, 2-bromo-pentane, 2-bromanylpentane, 2-bromopentane, 4-01-00-00312 beilstein handbook reference, ksc176i8j | |
| LGAJYTCRJPCZRJ-UHFFFAOYSA-N | |
| 2-bromopentane | |
| 7890 | |
| 90% |
Chemical Identifiers
| 107-81-3 | |
| 151.047 | |
| LGAJYTCRJPCZRJ-UHFFFAOYSA-N | |
| 7890 | |
| CCCC(C)Br |
| C5H11Br | |
| MFCD00000160 | |
| pentane, 2-bromo, 2-pentyl bromide, 1-methylbutyl bromide, +/--2-bromopentane, sec-amyl bromide, 2-bromo-pentane, 2-bromanylpentane, 2-bromopentane, 4-01-00-00312 beilstein handbook reference, ksc176i8j | |
| 2-bromopentane |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2-BROMOPENTANE
EINECSNumber : 203-521-3
RTECSNumber : RZ9800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only