missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromonicotinaldehyde, TCI America™

$24.44 - $70.28

Chemical Identifiers

CAS 128071-75-0
Molecular Formula C6H4BrNO
Molecular Weight (g/mol) 186.008
InChI Key GNFWMEFWZWXLIN-UHFFFAOYSA-N
Synonym 2-bromonicotinaldehyde, 2-bromo-3-formylpyridine, 2-bromo-3-pyridinecarboxaldehyde, 2-bromopyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 2-bromo, pubchem5057, acmc-1bypv, 2-bromo-3-formyl pyridine, bromo-3-pyridine carbaldehyde, bromo-3-pyridinecarboxaldehyde
PubChem CID 4714951
IUPAC Name 2-bromopyridine-3-carbaldehyde
SMILES C1=CC(=C(N=C1)Br)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B26251G View Documents TCI America
B26251G
1g Each for $24.44
 
B26255G View Documents TCI America
B26255G
5g Each for $70.28
 
Specifications

Chemical Identifiers

128071-75-0
186.008
2-bromonicotinaldehyde, 2-bromo-3-formylpyridine, 2-bromo-3-pyridinecarboxaldehyde, 2-bromopyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 2-bromo, pubchem5057, acmc-1bypv, 2-bromo-3-formyl pyridine, bromo-3-pyridine carbaldehyde, bromo-3-pyridinecarboxaldehyde
2-bromopyridine-3-carbaldehyde
C6H4BrNO
GNFWMEFWZWXLIN-UHFFFAOYSA-N
4714951
C1=CC(=C(N=C1)Br)C=O

Specifications

100°C
73°C
128071-75-0
2-bromonicotinaldehyde, 2-bromo-3-formylpyridine, 2-bromo-3-pyridinecarboxaldehyde, 2-bromopyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 2-bromo, pubchem5057, acmc-1bypv, 2-bromo-3-formyl pyridine, bromo-3-pyridine carbaldehyde, bromo-3-pyridinecarboxaldehyde
C1=CC(=C(N=C1)Br)C=O
186.008
186.01
≥98.0% (GC)
Undesignated
1g
C6H4BrNO
GNFWMEFWZWXLIN-UHFFFAOYSA-N
2-bromopyridine-3-carbaldehyde
4714951
Crystal-Powder at 20°C
2-Bromonicotinaldehyde
Safety and Handling

Safety and Handling

TSCA : No