missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromonaphthalene, 99%, Thermo Scientific Chemicals
$71.71 - $240.39
Chemical Identifiers
CAS | 580-13-2 |
---|---|
Molecular Formula | C10H7Br |
Molecular Weight (g/mol) | 207.07 |
MDL Number | MFCD00004051 |
InChI Key | APSMUYYLXZULMS-UHFFFAOYSA-N |
PubChem CID | 11372 |
IUPAC Name | 2-bromonaphthalene |
SMILES | BrC1=CC=C2C=CC=CC2=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC179710050
|
Thermo Scientific Chemicals
179710050 |
5 g | Glass bottle |
Each for $71.71
|
|
||||
AC179710250
|
Thermo Scientific Chemicals
179710250 |
25 g | Glass bottle |
Each for $240.39
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
580-13-2 | |
207.07 | |
APSMUYYLXZULMS-UHFFFAOYSA-N | |
2-bromonaphthalene |
C10H7Br | |
MFCD00004051 | |
11372 | |
BrC1=CC=C2C=CC=CC2=C1 |
Specifications
580-13-2 | |
Brown-Orange to Yellow | |
>110°C | |
98.5% min. (GC) | |
C10H7Br | |
MFCD00004051 | |
05,548 | |
naphthalene, 2-bromo, 2-naphthyl bromide, 2-bromo-naphthalene, beta-bromonaphthalene, beta-naphthyl bromide, .beta.-bromonaphthalene, unii-syu33i753n, naphthalene,2-bromo, .beta.-naphthyl bromide, pbn-2 | |
APSMUYYLXZULMS-UHFFFAOYSA-N | |
2-bromonaphthalene | |
11372 | |
99% | |
2-Bromonaphthalene |
53.0°C to 57.0°C | |
281.0°C to 282.0°C | |
Authentic | |
Glass bottle | |
C10H7Br | |
5 g | |
15,1437 | |
Solubility in water: slightly soluble. Other solubilities: soluble in 8 parts alcohol,ether,chloroform and,benzene | |
BrC1=CC=C2C=CC=CC2=C1 | |
207.07 | |
207.07 | |
Liquid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 209-452-5
RUO – Research Use Only