missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromonaphthalene, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 179711000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Bromonaphthalene | |
281.0°C to 282.0°C | |
Authentic | |
Glass bottle | |
C10H7Br | |
100g | |
15,1437 | |
Solubility in water: slightly soluble. Other solubilities: soluble in 8 parts alcohol,ether,chloroform and,benzene | |
BrC1=CC=C2C=CC=CC2=C1 | |
207.07 | |
207.07 |
580-13-2 | |
>110°C | |
98.5% min. (GC) | |
C10H7Br | |
MFCD00004051 | |
05,548 | |
naphthalene, 2-bromo, 2-naphthyl bromide, 2-bromo-naphthalene, beta-bromonaphthalene, beta-naphthyl bromide, .beta.-bromonaphthalene, unii-syu33i753n, naphthalene,2-bromo, .beta.-naphthyl bromide, pbn-2 | |
APSMUYYLXZULMS-UHFFFAOYSA-N | |
2-bromonaphthalene | |
11372 | |
99% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 209-452-5
RUO â Research Use Only