missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromonaphthalene, 98+%

Manufacturer:  Alfa Aesar™ A1530022

 View more versions of this product

Catalog No. AAA1530022



Description

Description

Applications
2-Bromonaphthalene is used as a raw material in the preparation of biaryls through Suzuki cross coupling reaction. Further, it plays a vital role in the preparation of dyes. It is also used to study potential tumorigenicity.

Solubility
Slightly soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

2-Bromonaphthalene
Odorless
53°C to 57°C
580-13-2
MFCD00004051
14,1426
APSMUYYLXZULMS-UHFFFAOYSA-N
2-bromonaphthalene
11372
≥98%
112°C (233°F)
281°C to 282°C
100g
C10H7Br
1858110
naphthalene, 2-bromo, 2-naphthyl bromide, 2-bromo-naphthalene, beta-bromonaphthalene, beta-naphthyl bromide, .beta.-bromonaphthalene, unii-syu33i753n, naphthalene,2-bromo, .beta.-naphthyl bromide, pbn-2
C1=CC=C2C=C(C=CC2=C1)Br
207.07
207.08

Chemical Identifiers

580-13-2
207.07
APSMUYYLXZULMS-UHFFFAOYSA-N
11372
C1=CC=C2C=C(C=CC2=C1)Br
C10H7Br
MFCD00004051
naphthalene, 2-bromo, 2-naphthyl bromide, 2-bromo-naphthalene, beta-bromonaphthalene, beta-naphthyl bromide, .beta.-bromonaphthalene, unii-syu33i753n, naphthalene,2-bromo, .beta.-naphthyl bromide, pbn-2
2-bromonaphthalene
Safety and Handling

Safety and Handling

GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P301+P312-P337+P313-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents