Learn More
2-(Bromomethyl)pyridine hydrobromide, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 370111000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-(Bromomethyl)pyridine hydrobromide | |
97.5 | |
Authentic | |
Glass bottle | |
BrCH2(C5H3N) | |
100g | |
JQDNCGRNPYKRAO-UHFFFAOYSA-N | |
2-(bromomethyl)pyridine;hydrobromide | |
2734720 | |
98% |
31106-82-8 | |
100.0 | |
98% | |
C6H6BrN·HBr | |
MFCD01863544 | |
2-bromomethyl pyridine hydrobromide, 2-picolyl bromide hydrobromide, 2-bromomethyl pyridine hbr, pyridine, 2-bromomethyl-, hydrobromide, 2-bromomethyl pyridine, bromide, c6h6brn.hbr, acmc-20aim5, ksc495g6r, 2-bromomethylpyridine hydrobromide, bromomethyl pyridine hydrobromide | |
C1=CC=NC(=C1)CBr.Br | |
252.94 | |
252.94 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Danger
RUO â Research Use Only