missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Bromomethyl)naphthalene, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC162560250

 View more versions of this product

Catalog No. AC162560250



Specifications

Specifications

2-(Bromomethyl)naphthalene
Authentic
213°C (100.0mmHg)
52°C to 57°C
939-26-4
100.0
C11H9Br
MFCD00004123
.beta.-bromomethyl naphthalene
C1=CC=C2C=C(C=CC2=C1)CBr
221.1
221.1
96%
>110°C
Glass bottle
Yellow to Beige
25g
95.0
95% min. (GC)
C10H7CH2Br
05,568
RUHJZSZTSCSTCC-UHFFFAOYSA-N
2-(bromomethyl)naphthalene
70320
Fine Crystalline Powder

Chemical Identifiers

939-26-4
221.1
RUHJZSZTSCSTCC-UHFFFAOYSA-N
70320
C1=CC=C2C=C(C=CC2=C1)CBr
C11H9Br
MFCD00004123
.beta.-bromomethyl naphthalene
2-(bromomethyl)naphthalene
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 213-359-5

SDS Documents