Learn More
2-(Bromomethyl)naphthalene, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2456822
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water (reacts), and chloroform.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Specifications
2-(Bromomethyl)naphthalene | |
51°C to 55°C | |
>110°C (230°F) | |
MFCD00004123 | |
UN3261 | |
2-bromomethyl naphthalene, naphthalene, 2-bromomethyl, 2-naphthylmethyl bromide, 2-naphthyl bromomethane, 2-bromomethyl napthalene, .beta.-bromomethyl naphthalene, 2-brommethyl naphthalen, zlchem 566, 2-menaphthyl bromide | |
RUHJZSZTSCSTCC-UHFFFAOYSA-N | |
2-(bromomethyl)naphthalene | |
70320 | |
96% |
939-26-4 | |
213°C (100 mmHg) | |
C11H9Br | |
100 g | |
636546 | |
Soluble in water (reacts),and chloroform. | |
BrCC1=CC=C2C=CC=CC2=C1 | |
221.10 | |
221.1 |
Chemical Identifiers
939-26-4 | |
221.10 | |
RUHJZSZTSCSTCC-UHFFFAOYSA-N | |
70320 | |
BrCC1=CC=C2C=CC=CC2=C1 |
C11H9Br | |
MFCD00004123 | |
2-bromomethyl naphthalene, naphthalene, 2-bromomethyl, 2-naphthylmethyl bromide, 2-naphthyl bromomethane, 2-bromomethyl napthalene, .beta.-bromomethyl naphthalene, 2-brommethyl naphthalen, zlchem 566, 2-menaphthyl bromide | |
2-(bromomethyl)naphthalene |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
H314-H317-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 213-359-5
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only