Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Bromomethyl)benzoic acid, 97%

Chemical Identifiers

Chemical Name or Material 2-(Bromomethyl)benzoic acid
CAS 7115-89-1
Molecular Formula C8H7BrO2
Formula Weight 215.05
MDL Number MFCD00090420
UN Number UN3261
Synonym alpha-Bromo-o-toluic acid; 2-Carboxybenzyl bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2892709

 
alfa aesar™
H2892709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2892718

 
alfa aesar™
H2892718
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 215.05
Chemical Name or Material 2-(Bromomethyl)benzoic acid
Assay 97%
Molecular Formula C8H7BrO2
MDL Number MFCD00090420
CAS 7115-89-1
UN Number UN3261
Melting Point ≈145°C
Synonym alpha-Bromo-o-toluic acid; 2-Carboxybenzyl bromide
Quantity 10g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Applications
2-(Bromomethyl)benzoic acid contains bi functional groups such as benzylic bromide and carboxylic acid are involved in nucleophilic reactions and esterification reactions respectively.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.