missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Bromomethyl)benzeneboronic acid pinacol ester, 98%

Manufacturer:  Alfa Aesar™ H53217MD

 View more versions of this product

Catalog No. AAH53217MD



Specifications

Specifications

2-(Bromomethyl)benzeneboronic acid pinacol ester
250mg
C13H18BBrO2
2-2-bromomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-bromomethylphenylboronic acid pinacol ester, 2-2-bromomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-bromomethyl benzeneboronic acid, pinacol ester, 2-bromomethylphenylboronic acid, pinacol ester, 2-bromomethyl benzeneboronic acid pinacol ester, 2-bromomethylphenyl boronic acid, pinacol ester, amtb373, 2-bromomethyl phenylboronic pinacol ester, 2-bromoethylphenylboronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2CBr
296.999
297
78°C to 80°C
377780-72-8
MFCD01632210
ROIXSNLOYHDYBP-UHFFFAOYSA-N
2-[2-(bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
3834834
98%

Chemical Identifiers

377780-72-8
296.999
ROIXSNLOYHDYBP-UHFFFAOYSA-N
3834834
B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2CBr
C13H18BBrO2
MFCD01632210
2-2-bromomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-bromomethylphenylboronic acid pinacol ester, 2-2-bromomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-bromomethyl benzeneboronic acid, pinacol ester, 2-bromomethylphenylboronic acid, pinacol ester, 2-bromomethyl benzeneboronic acid pinacol ester, 2-bromomethylphenyl boronic acid, pinacol ester, amtb373, 2-bromomethyl phenylboronic pinacol ester, 2-bromoethylphenylboronic acid pinacol ester
2-[2-(bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SDS Documents