Please login to your online account to display your discounted pricing

2-(Bromomethyl)-6-pyrrolidin-1-ylpyridine, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC38008DA

 View more versions of this product

Catalog No. CC38008DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 241.13
Quantity 1g
Assay 97%
CAS Max % 100.0
Packaging Amber Glass Bottle
CAS 869901-04-2
Assay Percent Range 97%
Molecular Formula C10H13BrN2
Chemical Name or Material 2-(Bromomethyl)-6-pyrrolidin-1-ylpyridine
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD09817465
CAS Min % 96.5
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material 2-(Bromomethyl)-6-pyrrolidin-1-ylpyridine
CAS 869901-04-2
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H13BrN2
Formula Weight 241.13
MDL Number MFCD09817465
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.

SDS & Safety Documents