Learn More
2-Bromomethyl-6-methylpyridine, 97%, Thermo Scientific Chemicals
$50.63 - $512.95
Chemical Identifiers
CAS | 68470-59-7 |
---|---|
Molecular Formula | C7H8BrN |
Molecular Weight (g/mol) | 186.05 |
MDL Number | MFCD04967229 |
InChI Key | WJFDCFHWFHCLIW-UHFFFAOYSA-N |
Synonym | 2-bromomethyl-6-methylpyridine, pyridine, 2-bromomethyl-6-methyl, 6-methyl-2-bromomethyl pyridine, 2-bromomethyl-6-methyl-pyridine, 6-bromomethyl-2-methylpyridine, pubchem22577, acmc-209ur7, 2-methyl-6-bromomethyl-pyridine, 2-bromomethyl-6-methyl pyridine |
PubChem CID | 12543733 |
IUPAC Name | 2-(bromomethyl)-6-methylpyridine |
SMILES | CC1=NC(CBr)=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH55438MD
|
Thermo Scientific Chemicals
H55438MD |
250 mg |
Each for $50.63
|
|
|||||
AAH5543803
|
Thermo Scientific Chemicals
H5543803 |
1 g |
Each for $143.96
|
|
|||||
AAH5543806
|
Thermo Scientific Chemicals
H5543806 |
5 g |
Each for $512.95
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
68470-59-7 | |
186.05 | |
WJFDCFHWFHCLIW-UHFFFAOYSA-N | |
12543733 | |
CC1=NC(CBr)=CC=C1 |
C7H8BrN | |
MFCD04967229 | |
2-bromomethyl-6-methylpyridine, pyridine, 2-bromomethyl-6-methyl, 6-methyl-2-bromomethyl pyridine, 2-bromomethyl-6-methyl-pyridine, 6-bromomethyl-2-methylpyridine, pubchem22577, acmc-209ur7, 2-methyl-6-bromomethyl-pyridine, 2-bromomethyl-6-methyl pyridine | |
2-(bromomethyl)-6-methylpyridine |
Specifications
68470-59-7 | |
C7H8BrN | |
250 mg | |
2-bromomethyl-6-methylpyridine, pyridine, 2-bromomethyl-6-methyl, 6-methyl-2-bromomethyl pyridine, 2-bromomethyl-6-methyl-pyridine, 6-bromomethyl-2-methylpyridine, pubchem22577, acmc-209ur7, 2-methyl-6-bromomethyl-pyridine, 2-bromomethyl-6-methyl pyridine | |
CC1=NC(CBr)=CC=C1 | |
186.05 | |
186.05 | |
2-Bromomethyl-6-methylpyridine |
38°C to 42°C | |
MFCD04967229 | |
Air sensitive | |
WJFDCFHWFHCLIW-UHFFFAOYSA-N | |
2-(bromomethyl)-6-methylpyridine | |
12543733 | |
97% |
Safety and Handling
GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302-H315-H318-H335
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only