missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromomethyl-6-methylpyridine, 97%

Manufacturer:  Alfa Aesar™ H55438MD

 View more versions of this product

Catalog No. AAH55438MD



Specifications

Specifications

2-Bromomethyl-6-methylpyridine
250mg
C7H8BrN
2-bromomethyl-6-methyl pyridine
CC1=NC(=CC=C1)CBr
186.052
186.05
38°C to 42°C
68470-59-7
MFCD04967229
WJFDCFHWFHCLIW-UHFFFAOYSA-N
2-(bromomethyl)-6-methylpyridine
12543733
97%

Chemical Identifiers

68470-59-7
186.052
WJFDCFHWFHCLIW-UHFFFAOYSA-N
12543733
CC1=NC(=CC=C1)CBr
C7H8BrN
MFCD04967229
2-bromomethyl-6-methyl pyridine
2-(bromomethyl)-6-methylpyridine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Recommended Storage : 2° to 8°C

Documents