Please login to your online account to display your discounted pricing

2-(Bromomethyl)-5-(trifluoromethyl)furan, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  MO00141DA

 View more versions of this product

Catalog No. MO00141DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 228.99
Quantity 1g
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
Health Hazard 1 GHS Signal Word: Danger
CAS 17515-77-4
Molecular Formula C6H4BrF3O
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Chemical Name or Material 2-(Bromomethyl)-5-(trifluoromethyl)furan
Assay Percent Range 97%
MDL Number MFCD03086219
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-(Bromomethyl)-5-(trifluoromethyl)furan
CAS 17515-77-4
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H4BrF3O
Formula Weight 228.99
MDL Number MFCD03086219
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents