missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromomethyl-4-fluorobenzonitrile, 98%

Manufacturer:  Alfa Aesar™ H6311003

 View more versions of this product

Catalog No. AAH6311003



Specifications

Specifications

2-Bromomethyl-4-fluorobenzonitrile
421552-12-7
MFCD08059542
2-cyano-5-fluorobenzyl bromide, 2-bromomethyl-4-fluorobenzonitrile, 2-cyano-5-fluorobenzylbromide, 2-cyano-5-fluoro benzyl bromide, 2-bromomethyl-4-fluoro-benzonitrile, 2-bromomethyl-4-fluorobenzenecarbonitrile, pubchem4906, acmc-1arut, ksc493s9t, 2-cyano-5-flurobenzyl bromide
C1=CC(=C(C=C1F)CBr)C#N
214.037
214.04
98%
1g
C8H5BrFN
UN2923
CHCAGFNTASDQFX-UHFFFAOYSA-N
2-(bromomethyl)-4-fluorobenzonitrile
21984990
Solid

Chemical Identifiers

421552-12-7
214.037
CHCAGFNTASDQFX-UHFFFAOYSA-N
21984990
C1=CC(=C(C=C1F)CBr)C#N
C8H5BrFN
MFCD08059542
2-cyano-5-fluorobenzyl bromide, 2-bromomethyl-4-fluorobenzonitrile, 2-cyano-5-fluorobenzylbromide, 2-cyano-5-fluoro benzyl bromide, 2-bromomethyl-4-fluoro-benzonitrile, 2-bromomethyl-4-fluorobenzenecarbonitrile, pubchem4906, acmc-1arut, ksc493s9t, 2-cyano-5-flurobenzyl bromide
2-(bromomethyl)-4-fluorobenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

SDS Documents