Please login to your online account to display your discounted pricing

2-(Bromomethyl)-2-(hydroxymethyl)-1,3-propanediol, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC319630010

 View more versions of this product

Catalog No. AC319630010

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥98.5%
Formula Weight 199.05
Quantity 1g
Color White to light beige
Melting Point 70° to 77°C
CAS Max % 100.0
Chemical Name or Material 2-(Bromomethyl)-2-(hydroxymethyl)-1, 3-propanediol
Assay Percent Range 99%
Molecular Formula C5H11BrO3
Linear Formula BrCH2C(CH2OH)3
MDL Number MFCD00135306
CAS 19184-65-7
CAS Min % 98.5
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material 2-(Bromomethyl)-2-(hydroxymethyl)-1, 3-propanediol
CAS 19184-65-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C5H11BrO3
Linear Formula BrCH2C(CH2OH)3
Formula Weight 199.05
MDL Number MFCD00135306