Please login to your online account to display your discounted pricing

2-(Bromomethyl)-2-(hydroxymethyl)-1,3-propanediol, 99%, ACROS Organics™

$93.66 - $93.66

Chemical Identifiers

Chemical Name or Material 2-(Bromomethyl)-2-(hydroxymethyl)-1, 3-propanediol
CAS 19184-65-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C5H11BrO3
Linear Formula BrCH2C(CH2OH)3
Formula Weight 199.05
MDL Number MFCD00135306
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC319630010

  SDS
thermo fisher scientific
AC319630010
Glass bottle 1g Each for $93.66

AC319630050

  SDS
thermo fisher scientific
AC319630050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥98.5%
Formula Weight 199.05
Quantity 1g
Color White to light beige
Melting Point 70° to 77°C
CAS Max % 100.0
Chemical Name or Material 2-(Bromomethyl)-2-(hydroxymethyl)-1, 3-propanediol
Assay Percent Range 99%
Molecular Formula C5H11BrO3
Linear Formula BrCH2C(CH2OH)3
MDL Number MFCD00135306
CAS 19184-65-7
CAS Min % 98.5
Infrared Spectrum Authentic
Packaging Glass bottle

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.