Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromomethyl-1,4-benzodioxane, 97%

$34.49 - $370.97

Chemical Identifiers

Chemical Name or Material 2-Bromomethyl-1,4-benzodioxane
CAS 2164-34-3
Molecular Formula C9H9BrO2
Formula Weight 229.07
MDL Number MFCD00044831
Beilstein 150154
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2214903

  SDS
alfa aesar™
B2214903
1g Each for $34.49

AAB2214906

  SDS
alfa aesar™
B2214906
5g Each for $92.81

AAB2214914

  SDS
alfa aesar™
B2214914
25g Each for $370.97
Description & Specifications

Specifications

Formula Weight 229.07
Chemical Name or Material 2-Bromomethyl-1,4-benzodioxane
Assay 97%
Molecular Formula C9H9BrO2
MDL Number MFCD00044831
CAS 2164-34-3
Beilstein 150154
Boiling Point 143° to 146°C (8mmHg)
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.