Please login to your online account to display your discounted pricing

2-(Bromomethyl)-1,3-benzothiazole, 95%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC06008DE

 View more versions of this product

Catalog No. CC06008DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 228.11
Quantity 5g
Assay 95%
Assay Percent Range 95%
CAS 106086-78-6
Molecular Formula C8H6BrNS
Packaging Amber Glass Bottle
Chemical Name or Material 2-(Bromomethyl)-1, 3-benzothiazole

Specifications

Chemical Name or Material 2-(Bromomethyl)-1, 3-benzothiazole
Assay 95%
Assay Percent Range 95%
Molecular Formula C8H6BrNS
Formula Weight 228.11