Learn More
2-Bromomesitylene, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1227730
Description
2-Bromomesitylene is a useful synthetic intermediate, Aromatics, Boron Derivatives, Catalyst. It is an intermediate used to prepare Hydrogen[4-[bis(2,4,6-trimethylphenyl)phosphino]-2,3,5,6-tetrafluorophenyl]hydrobis(2,3,4,5,6-pentafluorophenyl)borate (H714510) which is a frustrated phosphonium borate for metal-free catalytic hydrogenation of imines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromomesitylene is a useful synthetic intermediate, Aromatics, Boron Derivatives, Catalyst. It is an intermediate used to prepare Hydrogen[4-[bis(2,4,6-trimethylphenyl)phosphino]-2,3,5,6-tetrafluorophenyl]hydrobis(2,3,4,5,6-pentafluorophenyl)borate (H714510) which is a frustrated phosphonium borate for metal-free catalytic hydrogenation of imines.
Solubility
Insoluble in water(20°C) .
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with Strong oxidizing agents, excess heat.
Specifications
2-Bromomesitylene | |
2°C | |
224°C to 226°C | |
C9H11Br | |
MFCD00000073 | |
1907245 | |
2-bromomesitylene, 2,4,6-trimethylbromobenzene, mesityl bromide, bromomesitylene, benzene, 2-bromo-1,3,5-trimethyl, 2,4,6-trimethybromombenzene, mesitylene, 2-bromo, 2,4,6-trimethylphenyl bromide, 1-bromo-2,4,6-trimethylbenzene, 2-bromo-1,3,5-trimethyl-benzene | |
RRTLQRYOJOSPEA-UHFFFAOYSA-N | |
2-bromo-1,3,5-trimethylbenzene | |
68473 | |
99% |
576-83-0 | |
1.322 | |
96°C (204°F) | |
1.552 | |
250 g | |
Light sensitive | |
Insoluble in water(20°C) . | |
CC1=CC(=C(C(=C1)C)Br)C | |
199.091 | |
199.1 |
Chemical Identifiers
576-83-0 | |
199.091 | |
RRTLQRYOJOSPEA-UHFFFAOYSA-N | |
68473 | |
CC1=CC(=C(C(=C1)C)Br)C |
C9H11Br | |
MFCD00000073 | |
2-bromomesitylene, 2,4,6-trimethylbromobenzene, mesityl bromide, bromomesitylene, benzene, 2-bromo-1,3,5-trimethyl, 2,4,6-trimethybromombenzene, mesitylene, 2-bromo, 2,4,6-trimethylphenyl bromide, 1-bromo-2,4,6-trimethylbenzene, 2-bromo-1,3,5-trimethyl-benzene | |
2-bromo-1,3,5-trimethylbenzene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 209-405-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only