missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromomesitylene, 99%

Manufacturer:  Alfa Aesar™ A1227718*

 View more versions of this product

Catalog No. AAA1227718



Description

Description

Applications
2-Bromomesitylene is a useful synthetic intermediate, Aromatics, Boron Derivatives, Catalyst. It is an intermediate used to prepare Hydrogen[4-[bis(2,4,6-trimethylphenyl)phosphino]-2,3,5,6-tetrafluorophenyl]hydrobis(2,3,4,5,6-pentafluorophenyl)borate (H714510) which is a frustrated phosphonium borate for metal-free catalytic hydrogenation of imines.

Solubility
Insoluble in water(20°C) .

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with Strong oxidizing agents, excess heat.
Specifications

Specifications

2-Bromomesitylene
2°C
50g
99%
MFCD00000073
2-Bromo-1,3,5-trimethylbenzene; 2,4,6-Trimethylbromobenzene
CC1=CC(=C(C(=C1)C)Br)C
199.091
199.1
576-83-0
224°C to 226°C
96°C (204°F)
C9H11Br
1907245
RRTLQRYOJOSPEA-UHFFFAOYSA-N
2-bromo-1,3,5-trimethylbenzene
68473

Chemical Identifiers

576-83-0
199.091
RRTLQRYOJOSPEA-UHFFFAOYSA-N
68473
CC1=CC(=C(C(=C1)C)Br)C
C9H11Br
MFCD00000073
2-Bromo-1,3,5-trimethylbenzene; 2,4,6-Trimethylbromobenzene
2-bromo-1,3,5-trimethylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents