Please login to your online account to display your discounted pricing

2-Bromomalonaldehyde, 97+%, Acros Organics


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC397900010

 View more versions of this product

Catalog No. AC397900010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 150.95
Quantity 1g
Chemical Name or Material 2-Bromomalonaldehyde
Packaging Glass bottle
Assay Percent Range 97+%
Molecular Formula C3H3BrO2
CAS 2065-75-0

Specifications

Chemical Name or Material 2-Bromomalonaldehyde
Assay Percent Range 97+%
Molecular Formula C3H3BrO2
Formula Weight 150.95