missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromoisonicotinic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC437380010

 View more versions of this product

Catalog No. AC437380010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Bromoisonicotinic acid
Glass bottle
212.0°C to 222.0°C
66572-56-3
100.0
C6H4BrNO2
YBTKGKVQEXAYEM-UHFFFAOYSA-N
2-bromopyridine-4-carboxylic acid
222701
Crystals or Crystalline Powder
Authentic
White
1g
96.0
96% min. (HPLC)
2-bromisonicotins, 2-bromo-4-pyridine carboxylic acid, 2-bromo-4-pyridinecarboxylic acid, 2-bromo-isonicotinic acid, 2-bromoisonicotic acid, 2-bromoisonicotinic acid, 2-bromopyridine-4-carboxylicacid, 4-pyridinecarboxylic acid, 2-bromo, acmc-209nvt, pubchem9211
C1=CN=C(C=C1C(=O)O)Br
202.01
202.01
97%

Chemical Identifiers

66572-56-3
202.01
2-bromisonicotins, 2-bromo-4-pyridine carboxylic acid, 2-bromo-4-pyridinecarboxylic acid, 2-bromo-isonicotinic acid, 2-bromoisonicotic acid, 2-bromoisonicotinic acid, 2-bromopyridine-4-carboxylicacid, 4-pyridinecarboxylic acid, 2-bromo, acmc-209nvt, pubchem9211
2-bromopyridine-4-carboxylic acid
C6H4BrNO2
YBTKGKVQEXAYEM-UHFFFAOYSA-N
222701
C1=CN=C(C=C1C(=O)O)Br
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS Documents