Please login to your online account to display your discounted pricing

2-Bromoindene, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC384040050

 View more versions of this product

Catalog No. AC384040050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 195.06
Physical Form Low Melting Solid
Quantity 5g
Color Yellow to pale brown
Melting Point 36° to 39°C
CAS Max % 100.0
Chemical Name or Material 2-Bromoindene
CAS 10485-09-3
MDL Number MFCD06797863
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Assay Percent Range 99%
CAS Min % 98.5
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C9H7Br

Specifications

Chemical Name or Material 2-Bromoindene
CAS 10485-09-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H7Br
Formula Weight 195.06
MDL Number MFCD06797863
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents