missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromoindene, 99%, ACROS Organics™

$130.75 - $130.75

Chemical Identifiers

CAS 10485-09-3
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C9H7Br
Formula Weight 195.06
MDL Number MFCD06797863
Chemical Name or Material 2-Bromoindene
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC384040050 SDS Thermo Fisher Scientific
AC384040050
Glass bottle 5g Each for $130.75
Specifications Description & Specifications

Chemical Identifiers

10485-09-3
100.0
C9H7Br
MFCD06797863
≥98.5%
98.5
99%
195.06
2-Bromoindene

Specifications

10485-09-3
100.0
Glass bottle
5g
Low Melting Solid
C9H7Br
MFCD06797863
≥98.5%
98.5
Authentic
36° to 39°C
Yellow to pale brown
99%
195.06
2-Bromoindene

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.