missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromohexanoic Acid, TCI America™

$30.56 - $302.50

Chemical Identifiers

CAS 616-05-7
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.056
MDL Number MFCD00004218
InChI Key HZTPKMIMXLTOSK-UHFFFAOYSA-N
Synonym 2-bromocaproic acid, dl-2-bromohexanoic acid, hexanoic acid, 2-bromo, alpha-bromohexanoic acid, alpha-bromo-n-caproic acid, .alpha.-bromo-n-caproic acid, 2-bromo-hexanoic acid, .alpha.-bromocaproic acid, .alpha.-bromohexanoic acid, 2-bromo-n-hexanoic acid
PubChem CID 12013
IUPAC Name 2-bromohexanoic acid
SMILES CCCCC(C(=O)O)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B060125G View Documents TCI America
B060125G
25g Each for $30.56
 
B0601500G View Documents TCI America
B0601500G
500g Each for $302.50
 
Specifications

Chemical Identifiers

616-05-7
195.056
HZTPKMIMXLTOSK-UHFFFAOYSA-N
12013
CCCCC(C(=O)O)Br
C6H11BrO2
MFCD00004218
2-bromocaproic acid, dl-2-bromohexanoic acid, hexanoic acid, 2-bromo, alpha-bromohexanoic acid, alpha-bromo-n-caproic acid, .alpha.-bromo-n-caproic acid, 2-bromo-hexanoic acid, .alpha.-bromocaproic acid, .alpha.-bromohexanoic acid, 2-bromo-n-hexanoic acid
2-bromohexanoic acid

Specifications

240°C
25g
C6H11BrO2
3265
HZTPKMIMXLTOSK-UHFFFAOYSA-N
2-bromohexanoic acid
12013
Clear Liquid at 20°C
2-Bromohexanoic Acid
Yellow
616-05-7
MFCD00004218
2-bromocaproic acid, dl-2-bromohexanoic acid, hexanoic acid, 2-bromo, alpha-bromohexanoic acid, alpha-bromo-n-caproic acid, .alpha.-bromo-n-caproic acid, 2-bromo-hexanoic acid, .alpha.-bromocaproic acid, .alpha.-bromohexanoic acid, 2-bromo-n-hexanoic acid
CCCCC(C(=O)O)Br
195.056
195.06
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3249

TSCA : Yes