Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromohexane, 98%

Chemical Identifiers

Chemical Name or Material 2-Bromohexane
CAS 3377-86-4
Molecular Formula C6H13Br
Formula Weight 165.08
MDL Number MFCD00039183
UN Number UN1993
Beilstein 1731328
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0336414

  SDS
alfa aesar™
L0336414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 165.08
CAS 3377-86-4
Molecular Formula C6H13Br
Health Hazard 1 Warning
UN Number UN1993
Health Hazard 3 GHS P Statement
P261
Avoid breathing dust/fume/gas/mist/vapors/spray.
Flash Point 47°C (116°F)
MDL Number MFCD00039183
Assay 98%
Odor Pleasant
Beilstein 1731328
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Quantity 25g
TSCA TSCA
Boiling Point 146° to 147°C
Chemical Name or Material 2-Bromohexane

Applications
2-Bromohexane can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible in water.

Notes
Store away from strong bases. Incompatible with oxidizing agents and bases. Light-sensitive.