missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromofuran, 97%, Maybridge

Manufacturer:  Thermo Scientific CC02010EA

 View more versions of this product

Catalog No. CC02010EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Bromofuran, stabilized with Copper (0.1%), Furan (0.5%) and Sodium Bicarbonate (0.4%)
10g
96.5,0.05,0.45,0.35
95%
MFCD00216609
OYMCMWPHMPODNK-UHFFFAOYSA-N
2-bromofuran
2776190
97%
Amber Glass Bottle
584-12-3,7440-50-8,110-00-9,144-55-8
98.5,0.15,0.55,0.45
C4H3BrO
furan, 2-bromo, bromofuran, 2-bromo-furan, 2-furyl bromide, alpha-furyl bromide, 2-bromofurane, 5-bromofuran, pubchem6929, acmc-209m4d
C1=COC(=C1)Br
146.971
146.97

Chemical Identifiers

584-12-3
146.971
OYMCMWPHMPODNK-UHFFFAOYSA-N
2776190
C1=COC(=C1)Br
C4H3BrO
MFCD00216609
furan, 2-bromo, bromofuran, 2-bromo-furan, 2-furyl bromide, alpha-furyl bromide, 2-bromofurane, 5-bromofuran, pubchem6929, acmc-209m4d
2-bromofuran
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

Documents