Please login to your online account to display your discounted pricing

2-Bromofuran, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC02010DE

 View more versions of this product

Catalog No. CC02010DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 146.97
Quantity 5g
Assay 97%
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
CAS 584-12-3
Chemical Name or Material 2-Bromofuran, stabilized with Copper (0.1%), Furan (0.5%) and Sodium Bicarbonate (0.4%)
Molecular Formula C4H3BrO
Assay Percent Range 95%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Max % 0.55
CAS Min % 0.35
CAS Max % 0.15
MDL Number MFCD00216609
CAS 144-55-8
CAS Max % 0.45
CAS Min % 0.05
CAS Min % 0.45
CAS 7440-50-8
CAS 110-00-9
CAS Max % 98.5
CAS Min % 96.5
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 2-Bromofuran, stabilized with Copper (0.1%), Furan (0.5%) and Sodium Bicarbonate (0.4%)
CAS 584-12-3
CAS Min % 96.5
CAS Max % 98.5
CAS 7440-50-8
CAS Min % 0.05
CAS Max % 0.15
CAS 110-00-9
CAS Min % 0.45
CAS Max % 0.55
CAS 144-55-8
CAS Min % 0.35
CAS Max % 0.45
Assay 97%
Assay Percent Range 95%
Molecular Formula C4H3BrO
Formula Weight 146.97
MDL Number MFCD00216609
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents