Learn More
2-Bromofuran, 97%, Maybridge
$128.32 - $128.32
Chemical Identifiers
CAS | 584-12-3 |
---|---|
Molecular Formula | C4H3BrO |
Molecular Weight (g/mol) | 146.971 |
MDL Number | MFCD00216609 |
InChI Key | OYMCMWPHMPODNK-UHFFFAOYSA-N |
Synonym | 2-bromo-furan |
PubChem CID | 2776190 |
IUPAC Name | 2-bromofuran |
SMILES | C1=COC(=C1)Br |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC02010DA SDS |
Fisher Scientific Company
CC02010DA |
1g | Amber Glass Bottle |
Each for $128.32
|
|
CC02010DE SDS |
Fisher Scientific Company
CC02010DE |
5g | Amber Glass Bottle | N/A | N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC02010EA |
Fisher Scientific Company
CC02010EA |
10g | Amber Glass Bottle | N/A | N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
584-12-3 | |
146.971 | |
OYMCMWPHMPODNK-UHFFFAOYSA-N | |
2776190 | |
C1=COC(=C1)Br |
C4H3BrO | |
MFCD00216609 | |
2-bromo-furan | |
2-bromofuran |
Specifications
Amber Glass Bottle | |
584-12-3,7440-50-8,110-00-9,144-55-8 | |
98.5,0.15,0.55,0.45 | |
C4H3BrO | |
2-bromo-furan | |
C1=COC(=C1)Br | |
146.971 | |
146.97 | |
2-Bromofuran, stabilized with Copper (0.1%), Furan (0.5%) and Sodium Bicarbonate (0.4%) |
1g | |
96.5,0.05,0.45,0.35 | |
95% | |
MFCD00216609 | |
OYMCMWPHMPODNK-UHFFFAOYSA-N | |
2-bromofuran | |
2776190 | |
97% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning