Learn More
2-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 106990100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Bromofluorobenzene | |
1072-85-1 | |
100.0 | |
1.6000g/mL | |
43°C | |
99% | |
C6H4BrF | |
BrC6H4F | |
10 mL | |
1.6 | |
Solubility in water: insoluble in water | |
C1=CC=C(C(=C1)F)Br | |
175 | |
175 | |
Liquid |
99% | |
98.5 | |
Colorless to Yellow | |
155.0°C to 157.0°C | |
Authentic | |
Glass bottle | |
1.5327 to 1.5347 | |
MFCD00000282 | |
01,77 | |
2-bromofluorobenzene, o-bromofluorobenzene, 2-fluorobromobenzene, o-fluorobromobenzene, 2-bromfluorbenzen, benzene, 1-bromo-2-fluoro, 1-fluoro-2-bromobenzene, benzene, 1-fluoro-2-bromo, ofbb, benzene, bromofluoro | |
IPWBFGUBXWMIPR-UHFFFAOYSA-N | |
1-bromo-2-fluorobenzene | |
61259 | |
99% |
Chemical Identifiers
1072-85-1 | |
175 | |
IPWBFGUBXWMIPR-UHFFFAOYSA-N | |
61259 | |
C1=CC=C(C(=C1)F)Br |
C6H4BrF | |
MFCD00000282 | |
2-bromofluorobenzene, o-bromofluorobenzene, 2-fluorobromobenzene, o-fluorobromobenzene, 2-bromfluorbenzen, benzene, 1-bromo-2-fluoro, 1-fluoro-2-bromobenzene, benzene, 1-fluoro-2-bromo, ofbb, benzene, bromofluoro | |
1-bromo-2-fluorobenzene |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear pro
GHS Signal Word: Warning
EINECSNumber : 214-018-3
RTECSNumber : DA1050000
TSCA : TSCA
RUO – Research Use Only