missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromoethyleacetate, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC160090050

 View more versions of this product

Catalog No. AC160090050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Bromoethyl acetate
71°C
Glass bottle
160.0°C to 162.0°C
-13.8°C
927-68-4
100.0
C4H7BrO2
MFCD00000235
RGHQKFQZGLKBCF-UHFFFAOYSA-N
2-bromoethyl acetate
13564
Liquid
1.5100g/mL
Authentic
1.4547
Colorless to Yellow
5g
96.0
97%
CH3CO2CH2CH2Br
bromoethyl acetate, 1-acetoxy-2-bromoethane, 2-bromoethylacetate, ethanol, 2-bromo-, acetate, 1-bromo-2-acetoxyethane, acetic acid 2-bromoethyl ester, unii-nrz256o7qb, ethanol, 2-bromo-, 1-acetate, beta-bromoethyl acetate, nrz256o7qb
CC(=O)OCCBr
167.002
167
97%

Chemical Identifiers

927-68-4
167.002
RGHQKFQZGLKBCF-UHFFFAOYSA-N
13564
CC(=O)OCCBr
C4H7BrO2
MFCD00000235
bromoethyl acetate, 1-acetoxy-2-bromoethane, 2-bromoethylacetate, ethanol, 2-bromo-, acetate, 1-bromo-2-acetoxyethane, acetic acid 2-bromoethyl ester, unii-nrz256o7qb, ethanol, 2-bromo-, 1-acetate, beta-bromoethyl acetate, nrz256o7qb
2-bromoethyl acetate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents