Please login to your online account to display your discounted pricing

2-Bromoethyleacetate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Bromoethyl acetate
CAS 927-68-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H7BrO2
Linear Formula CH3CO2CH2CH2Br
Formula Weight 167
MDL Number MFCD00000235
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC160090050

  SDS
thermo fisher scientific
AC160090050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC160090250

  SDS
thermo fisher scientific
AC160090250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC160091000

  SDS
thermo fisher scientific
AC160091000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥96.0%
Formula Weight 167
Quantity 5g
Color Clear colorless to yellow
Density 1.5100g/mL
CAS Max % 100.0
CAS Min % 96.0
Packaging Glass bottle
Linear Formula CH3CO2CH2CH2Br
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Assay Percent Range 97%
Chemical Name or Material 2-Bromoethyl acetate
Molecular Formula C4H7BrO2
CAS 927-68-4
MDL Number MFCD00000235
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes serious eye irritation.