missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromoethyl acetate, 97%, Acros Organics

Manufacturer:  Thermo Fisher Scientific AC160090250

 View more versions of this product

Catalog No. AC160090250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Bromoethyl acetate
71°C
Glass bottle
1.51
Colorless to Yellow
25g
96.0
96% min. (GC)
CH3CO2CH2CH2Br
1-acetoxy-2-bromoethane
CC(=O)OCCBr
167
167
97%
1.5100g/mL
Authentic
1.4547
160.0°C to 162.0°C
-13.8°C
927-68-4
100.0
C4H7BrO2
MFCD00000235
RGHQKFQZGLKBCF-UHFFFAOYSA-N
2-bromoethyl acetate
13564
Liquid

Chemical Identifiers

927-68-4
167
RGHQKFQZGLKBCF-UHFFFAOYSA-N
13564
CC(=O)OCCBr
C4H7BrO2
MFCD00000235
1-acetoxy-2-bromoethane
2-bromoethyl acetate
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 213-159-8

SDS Documents