missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromoethylamine hydrobromide, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 106930025

Catalog No. AC106930025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Bromoethylamine hydrobromide
2576-47-8,107-09-5
100.0
98.5% min. (Argentometry)
C2H6BrN·HBr
MFCD00012886
04,134
Solubility in water: >500g/L (20°C)
C(CBr)N.Br
204.9
204.9
99%
98.5
Authentic
Plastic bottle
BrCH2CH2NH2·HBr
2.5kg
2-bromoethylamine hydrobromide, 2-bromoethanamine hydrobromide, 2-aminoethyl bromide hydrobromide, 2-bromoethylamine hbr, 2-bromoethylammonium bromide, ethanamine, 2-bromo-, hydrobromide, 2-bromoethan-1-amine hydrobromide, bromoethylamine hydrobromide, aminoethyl bromide hbr, ethylamine, hydrobromide
WJAXXWSZNSFVNG-UHFFFAOYSA-N
2-bromoethanamine;hydrobromide
2774217
99%
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.<br

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 219-924-2

RTECSNumber : KQ8225000

TSCA : TSCA

Recommended Storage : this product has a strong avidity for water

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only