missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2-Bromoethyl)trimethylammonium Bromide 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 152350250

 View more versions of this product

Catalog No. AC152350250



Description

Description

Phase transfer catalyst
Specifications

Specifications

(2-Bromoethyl)trimethylammonium bromide
Glass Bottle
240°C to 244°C
98%
97.5
98%
BrCH2CH2N(CH3)3Br
glycine linoleamide
CCCCCC=CCC=CCCCCCCCC(=O)NCC(=O)O
337.50
246.97
≥97.5%
Authentic
Beige to Brown
25g
2764-03-6
100.0
C20H35NO3
MFCD00011865
YCRHZEHWEYAHCO-HZJYTTRNSA-N
2-[[(9Z,12Z)-octadeca-9,12-dienoyl]amino]acetic acid
6433346
Powder or Chunks

Chemical Identifiers

2764-03-6
337.50
YCRHZEHWEYAHCO-HZJYTTRNSA-N
6433346
CCCCCC=CCC=CCCCCCCCC(=O)NCC(=O)O
C20H35NO3
MFCD00011865
glycine linoleamide
2-[[(9Z,12Z)-octadeca-9,12-dienoyl]amino]acetic acid
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Toxic if swallowed.
Causes skin irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 220-419-4

RTECSNumber : BO9481000

TSCA : TSCA

SDS Documents