missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromoethyl Phenyl Sulfide, TCI America™

$119.17 - $362.09

Chemical Identifiers

CAS 4837-01-8
Molecular Formula C8H9BrS
Molecular Weight (g/mol) 217.12
MDL Number MFCD00040826
InChI Key UEFBOQYLXLEJSM-UHFFFAOYSA-N
Synonym 2-bromoethyl phenyl sulfide, 2-bromoethyl thio benzene, 2-bromoethyl phenyl sulfane, 2-bromoethyl sulfanyl benzene, 2-bromoethylthiobenzene, benzene, 2-bromoethyl thio, 2-bromo-1-phenylthioethane, 2-bromoethyl thio benzene, acmc-1aoyt, 2-phenylthioethyl bromide
PubChem CID 78553
IUPAC Name 2-bromoethylsulfanylbenzene
SMILES C1=CC=C(C=C1)SCCBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B20855G View Documents TCI America
B20855G
5g Each for $119.17
 
B208525G View Documents TCI America
B208525G
25g Each for $362.09
 
Specifications

Chemical Identifiers

4837-01-8
217.12
UEFBOQYLXLEJSM-UHFFFAOYSA-N
78553
C1=CC=C(C=C1)SCCBr
C8H9BrS
MFCD00040826
2-bromoethyl phenyl sulfide, 2-bromoethyl thio benzene, 2-bromoethyl phenyl sulfane, 2-bromoethyl sulfanyl benzene, 2-bromoethylthiobenzene, benzene, 2-bromoethyl thio, 2-bromo-1-phenylthioethane, 2-bromoethyl thio benzene, acmc-1aoyt, 2-phenylthioethyl bromide
2-bromoethylsulfanylbenzene

Specifications

137°C
5g
C8H9BrS
2-bromoethyl phenyl sulfide, 2-bromoethyl thio benzene, 2-bromoethyl phenyl sulfane, 2-bromoethyl sulfanyl benzene, 2-bromoethylthiobenzene, benzene, 2-bromoethyl thio, 2-bromo-1-phenylthioethane, 2-bromoethyl thio benzene, acmc-1aoyt, 2-phenylthioethyl bromide
C1=CC=C(C=C1)SCCBr
217.12
217.12
≥97.0% (GC)
Yellow
4837-01-8
MFCD00040826
UEFBOQYLXLEJSM-UHFFFAOYSA-N
2-bromoethylsulfanylbenzene
78553
Clear Liquid at 20°C
2-Bromoethyl Phenyl Sulfide
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator