Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromoethyl phenyl sulfide, 97%

$102.05 - $404.82

Chemical Identifiers

Chemical Name or Material 2-Bromoethyl phenyl sulfide
CAS 4837-01-8
Molecular Formula C8H9BrS
Linear Formula C6H5SCH2CH2Br
Formula Weight 217.12
MDL Number MFCD00040826
Beilstein 2206188
Synonym 2-(Phenylthio)ethyl bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1100506

 
alfa aesar™
L1100506
5g Each for $102.05

AAL1100514

 
alfa aesar™
L1100514
25g Each for $404.82
Description & Specifications

Specifications

Formula Weight 217.12
Linear Formula C6H5SCH2CH2Br
Assay 97%
Molecular Formula C8H9BrS
MDL Number MFCD00040826
CAS 4837-01-8
Beilstein 2206188
Synonym 2-(Phenylthio)ethyl bromide
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Chemical Name or Material 2-Bromoethyl phenyl sulfide
Boiling Point 140° to 141°C (10mmHg)
Quantity 5g
Health Hazard 1 Warning

Applications
2-Bromoethyl phenyl sulfide, is used to make other chemicals as well as pharmaceutical intermediate. It is also used as a application intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.