missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromoethyl phenyl sulfide, 97%

Manufacturer:  Alfa Aesar™ L1100514

 View more versions of this product

Catalog No. AAL1100514



Description

Description

Applications
2-Bromoethyl phenyl sulfide, is used to make other chemicals as well as pharmaceutical intermediate. It is also used as a application intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Specifications

2-Bromoethyl phenyl sulfide
25g
C8H9BrS
MFCD00040826
2-bromoethyl phenyl sulfide, 2-bromoethyl thio benzene, 2-bromoethyl phenyl sulfane, 2-bromoethyl sulfanyl benzene, 2-bromoethylthiobenzene, benzene, 2-bromoethyl thio, 2-bromo-1-phenylthioethane, 2-bromoethyl thio benzene, acmc-1aoyt, 2-phenylthioethyl bromide
C1=CC=C(C=C1)SCCBr
217.12
217.12
140°C to 141°C (10mmHg)
4837-01-8
C6H5SCH2CH2Br
2206188
UEFBOQYLXLEJSM-UHFFFAOYSA-N
2-bromoethylsulfanylbenzene
78553
97%

Chemical Identifiers

4837-01-8
217.12
UEFBOQYLXLEJSM-UHFFFAOYSA-N
78553
C1=CC=C(C=C1)SCCBr
C8H9BrS
MFCD00040826
2-bromoethyl phenyl sulfide, 2-bromoethyl thio benzene, 2-bromoethyl phenyl sulfane, 2-bromoethyl sulfanyl benzene, 2-bromoethylthiobenzene, benzene, 2-bromoethyl thio, 2-bromo-1-phenylthioethane, 2-bromoethyl thio benzene, acmc-1aoyt, 2-phenylthioethyl bromide
2-bromoethylsulfanylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Documents