missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromoethyl phenyl sulfide, 97%

Manufacturer:  Alfa Aesar™ L1100506

 View more versions of this product

Catalog No. AAL1100506



Description

Description

Applications
2-Bromoethyl phenyl sulfide, is used to make other chemicals as well as pharmaceutical intermediate. It is also used as a application intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Specifications

2-Bromoethyl phenyl sulfide
5g
C12H18BNO2
MFCD00040826
2-2-phenylethyl-1,3,6,2-dioxazaborocane, 2-phenylethyl-1-boronic acid diethanolamine ester, 2-phenylethyl-1-boronic acid diethanolamine cyclic ester
B1(OCCNCCO1)CCC2=CC=CC=C2
219.09
217.12
140°C to 141°C (10mmHg)
4848-04-8
C6H5SCH2CH2Br
2206188
NYYNBCZSIYCXIZ-UHFFFAOYSA-N
2-(2-phenylethyl)-1,3,6,2-dioxazaborocane
24208882
97%

Chemical Identifiers

4848-04-8
219.09
NYYNBCZSIYCXIZ-UHFFFAOYSA-N
24208882
B1(OCCNCCO1)CCC2=CC=CC=C2
C12H18BNO2
MFCD00040826
2-2-phenylethyl-1,3,6,2-dioxazaborocane, 2-phenylethyl-1-boronic acid diethanolamine ester, 2-phenylethyl-1-boronic acid diethanolamine cyclic ester
2-(2-phenylethyl)-1,3,6,2-dioxazaborocane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Documents