Learn More
2-Bromoethyl methyl ether, 96%, stab. with sodium carbonate, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1537014
Description
2-Bromoethyl methyl ether has been used as starting reagent in the synthesis of 1-bromo-4-(2-methoxyethoxy)benzene1 and 1-bromo-4-[(2-methoxyethoxy)methyl]benzene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromoethyl methyl ether has been used as starting reagent in the synthesis of 1-bromo-4-(2-methoxyethoxy)benzene1 and 1-bromo-4-[(2-methoxyethoxy)methyl]benzene.
Solubility
Soluble in water.14,4 g/L at 25°C
Notes
Protect from heat. Incompatible with heat, strong bases and oxidizing agents. Store at 2-8°C.
Specifications
2-Bromoethyl methyl ether | |
6482-24-2 | |
109°C to 110°C | |
Recognizable | |
1.448 | |
25 g | |
2-bromoethyl methyl ether, 2-methoxyethyl bromide, ethane, 1-bromo-2-methoxy, ether, 2-bromoethyl methyl, methoxyethyl bromide, 2-bromoethylmethylether, bromoethyl methyl ether, 2-bromoethylmethyl ether, 2-bromoethylmethyl-ether, 2-bromo-1-methoxyethane | |
YZUPZGFPHUVJKC-UHFFFAOYSA-N | |
1-bromo-2-methoxyethane | |
80972 | |
96% |
stabilized with sodium carbonate | |
1.478 | |
28°C (82°F) | |
C3H7BrO | |
MFCD00000236 | |
UN3271 | |
Soluble in water.14,4g/L at 25°C | |
COCCBr | |
138.992 | |
139 |
Chemical Identifiers
6482-24-2 | |
138.992 | |
YZUPZGFPHUVJKC-UHFFFAOYSA-N | |
80972 | |
COCCBr |
C3H7BrO | |
MFCD00000236 | |
2-bromoethyl methyl ether, 2-methoxyethyl bromide, ethane, 1-bromo-2-methoxy, ether, 2-bromoethyl methyl, methoxyethyl bromide, 2-bromoethylmethylether, bromoethyl methyl ether, 2-bromoethylmethyl ether, 2-bromoethylmethyl-ether, 2-bromo-1-methoxyethane | |
1-bromo-2-methoxyethane |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber : 229-339-4
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only