Learn More
2-Bromoethyl methyl ether, 95%, Thermo Scientific Chemicals
$36.03 - $36.03
Chemical Identifiers
CAS | 6482-24-2 |
---|---|
Molecular Formula | C3H7BrO |
Molecular Weight (g/mol) | 138.99 |
MDL Number | MFCD00000236 |
InChI Key | YZUPZGFPHUVJKC-UHFFFAOYSA-N |
Synonym | 2-bromoethyl methyl ether, 2-methoxyethyl bromide, ethane, 1-bromo-2-methoxy, ether, 2-bromoethyl methyl, methoxyethyl bromide, 2-bromoethylmethylether, bromoethyl methyl ether, 2-bromoethylmethyl ether, 2-bromoethylmethyl-ether, 2-bromo-1-methoxyethane |
PubChem CID | 80972 |
IUPAC Name | 1-bromo-2-methoxyethane |
SMILES | COCCBr |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC402980050
|
Thermo Scientific Chemicals
402980050 |
5 g | Glass bottle |
Each for $36.03
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6482-24-2 | |
138.99 | |
YZUPZGFPHUVJKC-UHFFFAOYSA-N | |
80972 | |
COCCBr |
C3H7BrO | |
MFCD00000236 | |
2-bromoethyl methyl ether, 2-methoxyethyl bromide, ethane, 1-bromo-2-methoxy, ether, 2-bromoethyl methyl, methoxyethyl bromide, 2-bromoethylmethylether, bromoethyl methyl ether, 2-bromoethylmethyl ether, 2-bromoethylmethyl-ether, 2-bromo-1-methoxyethane | |
1-bromo-2-methoxyethane |
Specifications
6482-24-2 | |
100.0 | |
1.4600g/mL | |
28°C | |
94% min. (GC) | |
C3H7BrO | |
BrCH2CH2OCH3 | |
5 g | |
2-bromoethyl methyl ether, 2-methoxyethyl bromide, ethane, 1-bromo-2-methoxy, ether, 2-bromoethyl methyl, methoxyethyl bromide, 2-bromoethylmethylether, bromoethyl methyl ether, 2-bromoethylmethyl ether, 2-bromoethylmethyl-ether, 2-bromo-1-methoxyethane | |
YZUPZGFPHUVJKC-UHFFFAOYSA-N | |
1-bromo-2-methoxyethane | |
80972 | |
95% | |
2-Bromoethyl methyl ether |
94.0 | |
Colorless to Yellow | |
107.0°C to 110.0°C | |
Authentic | |
Glass bottle | |
1.4460 to 1.4500 | |
MFCD00000236 | |
1.46 | |
Solubility in water: 14.4g/L (25°C) | |
COCCBr | |
138.99 | |
138.99 | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap a
GHS Signal Word: Warning
EINECSNumber : 229-339-4
Recommended Storage : Product darkens during storage
RUO – Research Use Only