missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromoethyl Ethyl Ether, TCI America™

$51.95 - $481.27

Chemical Identifiers

CAS 592-55-2
Molecular Formula C4H9BrO
Molecular Weight (g/mol) 153.019
MDL Number MFCD00000237
InChI Key MMYKTRPLXXWLBC-UHFFFAOYSA-N
Synonym 2-bromoethyl ethyl ether, 2-ethoxyethyl bromide, 2-bromoethoxyethane, ethane, 1-bromo-2-ethoxy, 2-bromodiethyl ether, 1-ethoxy-2-bromoethane, 1-bromo-2-ethoxy-ethane, ether, 2-bromoethyl ethyl, 2-bromoethylethylether, 1-bromo-3-oxapentane
PubChem CID 61141
IUPAC Name 1-bromo-2-ethoxyethane
SMILES CCOCCBr
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B059525G View Documents TCI America
B059525G
25g Each for $51.95
 
B0595500G View Documents TCI America
B0595500G
500g Each for $481.27
 
B0595100G View Documents TCI America
B0595100G
100g Each for $137.50
 
Specifications

Chemical Identifiers

592-55-2
153.019
MMYKTRPLXXWLBC-UHFFFAOYSA-N
61141
CCOCCBr
C4H9BrO
MFCD00000237
2-bromoethyl ethyl ether, 2-ethoxyethyl bromide, 2-bromoethoxyethane, ethane, 1-bromo-2-ethoxy, 2-bromodiethyl ether, 1-ethoxy-2-bromoethane, 1-bromo-2-ethoxy-ethane, ether, 2-bromoethyl ethyl, 2-bromoethylethylether, 1-bromo-3-oxapentane
1-bromo-2-ethoxyethane

Specifications

126°C
25g
C4H9BrO
2340
MMYKTRPLXXWLBC-UHFFFAOYSA-N
1-bromo-2-ethoxyethane
61141
Clear Liquid at 20°C
2-Bromoethyl Ethyl Ether
Yellow
592-55-2
MFCD00000237
2-bromoethyl ethyl ether, 2-ethoxyethyl bromide, 2-bromoethoxyethane, ethane, 1-bromo-2-ethoxy, 2-bromodiethyl ether, 1-ethoxy-2-bromoethane, 1-bromo-2-ethoxy-ethane, ether, 2-bromoethyl ethyl, 2-bromoethylethylether, 1-bromo-3-oxapentane
CCOCCBr
153.019
153.02
≥95.0% (GC)
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Freezer