Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromoethyl ethyl ether, 95%, stab. with 0.5% Potassium carbonate

Chemical Identifiers

Chemical Name or Material 2-Bromoethyl ethyl ether
CAS 592-55-2
Name Note stabilized with 0.5% Potassium carbonate
Molecular Formula C4H9BrO
Linear Formula BrCH2CH2OCH2CH3
Formula Weight 153.02
MDL Number MFCD00000237
UN Number UN2340
Beilstein 1731362
Synonym 2-Ethoxyethyl bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0403414

  SDS
alfa aesar™
L0403414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0403422

  SDS
alfa aesar™
L0403422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 153.02
Chemical Name or Material 2-Bromoethyl ethyl ether
Molecular Formula C4H9BrO
MDL Number MFCD00000237
Health Hazard 1 Danger
Name Note stabilized with 0.5% Potassium carbonate
Assay 95%
CAS 592-55-2
Beilstein 1731362
Synonym 2-Ethoxyethyl bromide
Boiling Point 127° to 128°C
Quantity 25g
Health Hazard 2 GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage −30° to −10°C
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2-BROMOETHYL ETHYL ETHER
TSCA TSCA
Linear Formula BrCH2CH2OCH2CH3
UN Number UN2340
Flash Point 21°C (69°F)

Applications
2-Bromoethyl ethyl ether is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.

Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

−30° to −10°C