Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromoethyl ethyl ether, 95%, stab. with 0.5% Potassium carbonate


Quantity 100g

Manufacturer: alfa aesar™  L04034-22

 View more versions of this product

Catalog No. AAL0403422

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Boiling Point 127° to 128°C
Quantity 100g
Flash Point 21°C (69°F)
Chemical Name or Material 2-Bromoethyl ethyl ether
Name Note stabilized with 0.5% Potassium carbonate
CAS 592-55-2
Assay 95%
Molecular Formula C4H9BrO
Linear Formula BrCH2CH2OCH2CH3
Formula Weight 153.02g/mol
MDL Number MFCD00000237
UN Number UN2340
Beilstein 1731362
Synonym 2-Ethoxyethyl bromide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage −30° to −10°C
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2-BROMOETHYL ETHYL ETHER
TSCA TSCA

Specifications

Chemical Name or Material 2-Bromoethyl ethyl ether
CAS 592-55-2
Name Note stabilized with 0.5% Potassium carbonate
Molecular Formula C4H9BrO
Linear Formula BrCH2CH2OCH2CH3
Formula Weight 153.02g/mol
MDL Number MFCD00000237
UN Number UN2340
Beilstein 1731362
Synonym 2-Ethoxyethyl bromide

Applications
2-Bromoethyl ethyl ether is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.

Accessories

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

−30° to −10°C