Please login to your online account to display your discounted pricing

(2-Bromoethyl)benzene, 98%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: fisher scientific acros  106945000

 View more versions of this product

Catalog No. AC106945000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 185.06
Quantity 500mL
Color Clear colorless to pale yellow
EINECS Number 203-130-8
TSCA TSCA
RTECS Number CY9032000
CAS Max % 100.0
Synonym β-Bromophenylethane
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C8H9Br
Linear Formula C6H5(CH2)2Br
MDL Number MFCD00000240
CAS 103-63-9
CAS Min % 97.5
Chemical Name or Material (2-Bromoethyl)benzene
Name Note 98%
Assay Percent Range 98%
Refractive Index 1.5553 to 1.5573 (20°C, 589nm)
Water 0.05% max.
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.

Specifications

Chemical Name or Material (2-Bromoethyl)benzene
Name Note 98%
CAS 103-63-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H9Br
Linear Formula C6H5(CH2)2Br
Formula Weight 185.06
MDL Number MFCD00000240
Synonym β-Bromophenylethane
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.

SDS & Safety Documents